Would you like to log in again?
Overhead Flow Table
Supplier: DESK LABS
470200-900 1
470200-900KT 343.75 USD InStock
470200-900 470018-268
Overhead Flow Table
Models Geology Models
Demonstrate Earth Science Concepts in the Classroom
Simulate over 100 real-world water, air, and magma events through your overhead projector or hands-on labs. This kit includes 16 different plates that allow you to demonstrate everything from cloud formation to volcanic eruptions. This self-contained unit takes the place of a ripple tank, sand table or wave tank—allowing you to easily illustrate difficult earth and physical science concepts on the overhead. It can also be used as a wind tunnel.
This versatile demonstration is easy to clean and store.
Kit may be used on a classroom table, with an overhead projector or with a digital document camera projection system.
Ordering Information: Kit includes unit (16 x 16 x 2") chambered tray, water pump, tubing, fittings, event plates, eyedropper and teacher's guide with instructions. Kit does not include projection system, food coloring or rheoscopic fluid.
Ordering information: Kit includes unit (16 x 16 x 2") chambered tray, water pump, tubing, fittings, event plates, eyedropper and teacher's guide with instructions. Kit does not include projection system, food coloring or rheoscopic fluid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty