Would you like to log in again?
Fundamantals of Physics Ballistic Pendulum
Supplier: VWR International
BLP-101
470018-434EA 93.85 USD InStock
470018-434
Fundamantals of Physics Ballistic Pendulum
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
The Ballistic Pendulum is spring-loaded for easy investigations.
  • Three Firing Positions
  • Pendulum Catch
  • Spring-Loaded Launcher
  • Horizontal Launcher
  • Students Research Conservation of Momentum and Projectiles
Students can research the conservation of momentum and projectiles with this spring-loaded launcher, which features three firing positions and a pendulum catch. Users can quickly attach a photogate to measure the actual velocity of the marble. It also works as horizontal launcher.
A photogate is available separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty