Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Tangent Height Gauge
Supplier: PECO SALES INC SE
36953
470018-570EA 45 USD InStock
470018-570
Tangent Height Gauge
Educational Materials Environmental Educational Materials Environmental Learning Activities
Teach Students to Quickly Determine Tree Heights
This mirror sighting gauge includes easy to follow instructions embossed directly onto its rugged plastic shell. The instructions detail the gauge’s short four step measuring process as well as height-finding formulas.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty