Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Field Guide for Soil and Stratigraphic Analysis Chart
Supplier: MIDWEST GEOSCIENCES GROUP
FG SOIL
470018-588EA 29.99 USD InStock
470018-588
Field Guide for Soil and Stratigraphic Analysis Chart
Charts
Enhance Your Soil and Sediment Descriptions
Improve the accuracy of your boring logs and classification of soils in the field using this field guide. The front side of the text includes a flow chart listing key descriptive elements necessary for characterizing sedimentary sequences on boring logs. Common terms from geologic, engineering, and soil science disciplines are also used to help prepare meaningful boring logs that are clear and concise. The back side includes an easy-to-use flow chart that takes the mystery out of using the Unified Soil Classification System and assigning a group name and USCS symbol in the field. Guide is printed on durable, double-sided, plastic-coated paper. Size: 8 1/2" x 11".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty