Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
USDA Soil Texturing Field Flow Chart
Supplier: MIDWEST GEOSCIENCES GROUP
USDA FC
470018-592EA 29.99 USD InStock
470018-592
USDA Soil Texturing Field Flow Chart
Charts
Preparation for Work in the Field
For assisting with soil texturing procedures and assigning a texture class using the USDA texture triangle, this chart uses step-wise procedures for estimating sand, silt, and clay content. The ribbon method is implemented to estimate the fine-grained fraction followed by placing the fraction within the texture triangle to determine Texture Class (ex: sandy loam). The accompanying guide provides instruction for determining subclass (ex: fine sandy loam) and assigning textural class modifiers for rock fragments greater than 2mm (ex: gravelly silt loam) and organic compositional textural modifiers (ex: peaty sandy clay loam). Guide is printed on durable, double-sided, plastic-coated paper.
Size: 8 1/2" x 11".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty