Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ward's® Ruler, 6"

Supplier: ACME UNITED CORP SE
18
470018-910BX 2.35 USD InStock
470018-910
Ward's® Ruler, 6"
Rulers
Allows You to See Your Work

  • Clear, flexible plastic
  • Graduated in 1 mm and 1/16" increments

In addition to measuring in millimeters and inches, this ruler also includes a list of metric, English, and Avoirdupois weights, plus cork and rubber stopper sizes on the back. Plastic, graduated in 1 mm and 1/16" increments.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty