Would you like to log in again?
Sharp-Point Surgical Scissors
Supplier: Dunrite
140981 140980 140982
470106-340EA 7.9 USD InStock
470106-340 470018-940 470018-942
Sharp-Point Surgical Scissors
Scissors
Start your cuts easily.
  • Dissect without a scalpel
  • Handles rounded for ambidextrous use
  • Available in two sizes
Sharp-pointed, straight surgical scissors allow the user to insert blades into tissue to start the cut. Larger scissors provide greater leverage for tough cutting; shorter scissors for finer work. Stainless steel.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty