Would you like to log in again?
Blunt-Point Surgical Scissors
Supplier: Dunrite
140985
470018-946BX 171 USD InStock
470018-946
Blunt-Point Surgical Scissors
Scissors
When working with particularly delicate specimens, the blunt tips of these straight scissors reduce the chance of tearing, damaging, or puncturing tissue adjacent to the cut. Stainless steel.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty