Would you like to log in again?
Rectangular Glass Coverslips
Supplier: C & A SCIENTIFIC CO INC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
94-2440 94-22402 94-2240
470019-010PK 7.1 USD InStock
470019-010 470145-746 470019-008
Rectangular Glass Coverslips
Cover Glasses
Traditional rectangular coverslip style
  • Quality slide making supplies
  • Traditional coverslip type
Traditional style coverslips to use when making slides in the laboratory.
#1 Thickness: 0.15 mm thick. Approximately 75–100 coverslips per ounce.
#2 Thickness: 0.2 mm thick. Approximately 50-75 coverslips per ounce.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty