Would you like to log in again?
Complete Blood Pressure Kit
Supplier: HEALTH SMART SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
04-176-021
470019-078CS 1313.85 USD InStock
470019-078
Complete Blood Pressure Kit
Blood Pressure Monitors
Everything needed to measure blood pressure and pulse is included in this economical kit.
  • Accurate to ±3 mm Hg
  • Zippered, Vinyl Storage Case
  • Included Recording Chart for Individual Record Keeping
It includes a stethoscope and a zippered vinyl storage case with instructions and record chart.
The provided aneroid sphygmomanometer, which features a standard-size D-ring cuff with a Velcro closure, is accurate to ±3 mm Hg.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty