Would you like to log in again?
Hand Dynomometer, Vernier Software & Tech SE
Supplier: VERNIER SOFTWARE & TECH SE
HD-BTA
470019-118EA 123.99 USD InStock
470019-118
Hand Dynomometer, Vernier Software & Tech SE
Dynamometers
Measure grip strength and pinch strength.
  • Measurements can be made in Newtons or kilograms force
The strain-gage based isometric dynamometer can be used to measure grip strength and to perform muscle fatigue studies. The Hand Dynamometer can be used alone or in combination with EMG recordings for detailed studies of muscular activity.
Power: 7 mA at 5VDC
Safety range (maximum force without damage done to sensor): 0 to 850 N
For educational use only
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty