Would you like to log in again?
GSR/Temp 2X Biofeedback System
Supplier: THOUGHT TECHNOLOGY, LTD
T2120
470019-156EA 282.15 USD InStock
470019-156
GSR/Temp 2X Biofeedback System
Educational Materials Biology Educational Materials Animal Biology Learning Activites
See and Hear How You Control Your Body’s Reaction to Stress
This device correlates the relationship of stress to galvanic skin resistance (GSR), which is controlled by the sympathetic nervous system. It translates tiny tension-related changes in your skin pores into a measurable signal that can be seen on a meter, or heard with earphones (both included). This system also allows you to visualize the changes in skin temperature that occur as a result of stress. It comes with the GSR monitor, temperature sensor, dual sensitivity meter of visual feedback, biofeedback relaxation training cassette, one 9 V battery, and an instruction booklet, all packed in a carrying case. Size of monitor: 4" x 3" x 1 3/4".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty