Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Ward's® Introductory Electrophoresis Hardware Systems
Supplier: Avantor
WARD470014-626 WARD470004-096 WARD470019-166 WARD470004-094 WARD470004-092 WARD470024-008
470014-626EA 102 USD InStock
470014-626 470024-008 470019-166 470004-094 470004-096 470004-092
Ward's® Introductory Electrophoresis Hardware Systems
Educational Materials Chemistry Educational Materials Chromatography Learning Activities
Battery-powered electrophoresis.
  • Battery-powered gel boxes don't require a pricey power supply
  • Designed for the first-time user
  • Gel boxes run on low-voltage
Designed for the first-time user, the complete system comes with all the hardware you need to run electrophoresis tests. It safely runs on low-voltage batteries, which provide enough power to run up to ten gels.
Gel run time: about 120 minutes on five batteries, 60 minutes on ten batteries.
Ordering information: Each set includes an electrophoresis chamber with high-conductivity platinum electrodes that won’t corrode, gel casting tray, six-well comb, patch cords, and instructions. Purchase an individual set, or as a lab pack of five.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty