Would you like to log in again?
Clear 360° Goniometer
Supplier: Fabrication Enterprises
12-1000
470019-280CS 498.75 USD InStock
470019-280
Clear 360° Goniometer
Goniometers
Clear Plastic Permits Easy Observation
This goniometer is marked in 1° increments and has three scales calibrated to be used with the International Standards of Measurement. One arm has a linear scale in both inches and centimeters to allow accurate measurement of the joint’s axis and range of motion.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty