Would you like to log in again?
Ward's® Mouse Maze
Supplier: Dynalon
8551-2663
470019-324EA 669.99 USD InStock
470019-324
Ward's® Mouse Maze
Educational Classroom Kits and Activities
Removable walls let you change the configuration of the maze.
  • Perfect for studying the behavior of rodents and other small animals
This maze with removable walls allows students to set up any pattern they wish. The clear acrylic cover allows students to observe the animals progress without any danger of escape. Made of polypropylene, this durable maze is ideal for use with all age groups and is perfect for science fair students for years to come. The maze may also be put into a dishwasher or autoclave to remove trace smells and pheromones left behind.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty