Would you like to log in again?
Rectangular Supports
Supplier: Shiv Dial Sud & Sons
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CH12321/2 H-21232 SSB4X6-CI
470019-494EA 24.95 USD InStock
470019-494 470019-496 470019-526
Rectangular Supports
Support Stands
Tip-free stability for experiments.
  • Glassware can sit directly on base
  • Rods are zinc-plated steel
Heavy cast iron bases with black enamel finish. Includes a removable zinc-plated steel rod.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty