Would you like to log in again?
Thin Stem Pipettes
Supplier: AQUAPHOENIX SCIENTIFIC INC. SE
4-424C 4-424L
470019-808PK 37.4 USD InStock
470019-808 470019-810
Thin Stem Pipettes
Pipettes Pasteur/Transfer Pipettes
Versatile pipettes provide consistent drop size.
  • Chemically inert and non-toxic
  • Can be stretched to form micropipettes and other micro lab ware
These pipettes serve as ideal micro-reagent vials that can be heat-sealed and frozen.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty