Would you like to log in again?
ID Tents with Cut-Out Scale
Supplier: FORENSICS SOURCE
IDTC-A26Y
470020-128KT 163.85 USD InStock
470020-128
ID Tents with Cut-Out Scale
Educational Materials Forensics Educational Materials Forensics Supplies
Simultaneously Mark and Measure Evidence for Photography
  • Built-in Scales
  • Reusable
Versatile and valuable, these ID tents will save time and effort in collecting field evidence. Each tent has three surfaces; two vertical surfaces with A to Z markers, and a third horizontal surface with a 1 3/4" square cut-out, featuring a metric scale and photo cross-hairs. The design allows for multiple photo angles — whether perpendicular, wall photos, or depth measurement of holes and crevices. Each durable tent is stackable for convenient storage and contains holes for mounting to walls or holding flag markers. Size: 3" x 3". Set of 26.
Note: This product is available to US customers only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty