Would you like to log in again?
Tex Ultra-Light Magnetic Applicator
Supplier: FORENSICS SOURCE
1-0153
470020-204EA 18.15 USD InStock
470020-204
Tex Ultra-Light Magnetic Applicator
Educational Materials Forensics Educational Materials Forensics Supplies
Designed by Retired Houston Police Officer
  • Disposable
  • Tidy Application
Prevent contamination from surface to surface, from your brush to your kit, and from your brush to your jar of powder with this disposable magnetic brush and five disposable covers. Designed for use at hazardous crime scenes and to avoid cross contamination, the brushes allow you to continue working after a brush becomes contaminated, by simply putting on a new cover. Especially useful when dusting at a scene containing biohazards or other contaminants, these brushes can be disposed of after use. The soft plastic tip and powerful magnet allow the brush to float over surfaces on a cushion of powder with minimal touching of the tip to the surface. And, because the magnet stays engaged during use, you can use it in any position — even upside down.
Available to U.S. customers only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty