Would you like to log in again?
Ward's® Drag Sled
Supplier: INDUSTRIAL SUPPORT INC.
156268
470020-256EA 162.1 USD InStock
470020-256
Ward's® Drag Sled
Educational Materials Forensics Educational Materials Crime Scene Evidence Learning Activities
Apply Coefficient of Friction Principles to Your Forensics Lessons
Determine the amount of force necessary to create different tire skid marks. Taking into account the composition of the ground or roadway surface, and the weight of the vehicle, this revolutionary sled establishes the force that created specific tire skid marks. Made of durable plastic, it features a tire tread mounted on the bottom surface, a secured metal eye bolt, and 15 lbs. of weight included in the container, The external and internal trays enable you to organize and transport your extra accident investigation supplie in the field. Size: 15"L x 7 1/2"W x 9"H.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty