Would you like to log in again?
Fuming Hot Plate
Supplier: SIRCHIE
FHP100
470020-328EA 28.95 USD InStock
470020-328
Fuming Hot Plate
Educational Materials Forensics Educational Materials Forensics Supplies
Accelerate the Development of Cyanoacrylate
Simply place this hot plate inside your fuming chamber and add 5-10 drops of cyanoacrylate into the aluminum tray. Prints will then develop in a matter of minutes, saving hours of lab time. 100 VAC.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty