Would you like to log in again?
BIO-FOAM® Impression Foam
Supplier: FORENSICS SOURCE
4-1120
470020-454EA 19.45 USD InStock
470020-454
BIO-FOAM® Impression Foam
Educational Materials Forensics Educational Materials Forensics Supplies
A Clean, Quick, Easy Way to Make Footprint Impressions
Follow in the footsteps of actual forensic detectives with this professional field technique. Press any footwear into the blue BIO-FOAM®, and after removal, the foam retains a detailed footprint impression, which can be stored in the original cardboard container. For further study, a cast can be made of foam impressions by coating with hair spray and pouring in a prepared casting material (available separately). Designed for one use per BIO-FOAM®. Size: 14"L x 6"W x 2 1/4"H
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty