Would you like to log in again?
Disposable Impression Trays
Supplier: FRANKLIN DENTAL SUPPLY
IT-7004 IT-7003
470020-458PK 17.5 USD InStock
470020-458 470020-462
Disposable Impression Trays
Educational Materials Forensics Educational Materials Forensics Supplies
Inexpensive Recreations for Labwork
  • Autoclavable
  • Easy to Clean
Cast impressions of teeth to identify placement and study bite marks for forensic odontology lessons. This set of disposable dental impression trays is autoclavable and available for upper and lower jaw impressions. Set
of 12.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty