Would you like to log in again?
Flask Form Electroscope
Supplier: G.S.C. INTERNATIONAL, INC.
4-50150
470020-542EA 27.1 USD InStock
470020-542
Flask Form Electroscope
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Simple and Inexpensive
Use this compact apparatus contained in a 250 mL Pyrex flask. A nickel-plated rod with two aluminum leaves on one end fits into a rubber stopper in the neck of the flask. Charge the rod, and the leaves move apart.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty