Would you like to log in again?
Bar and Gauge Apparatus
Supplier: VWR International
PH0378A
470020-568EA 32 USD InStock
470020-568
Bar and Gauge Apparatus
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
A new way to demonstrate thermal expansion in a metal.
  • An alternative to the classic ball-and-ring thermal expansion apparatus
The bar and gauge allows learners to see how much the metal expands in both length and diameter. The bar, which fits closely into the gauge - either lengthwise or through the holes - will be noticeably longer once heated. Your hands are kept safe by a 8.25" handle with a wooden grip. Transform the traditional "magic trick" discrepant event into a real learning experience without losing any of the "WOW" factor.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty