Would you like to log in again?
Equilateral Prism
Supplier: United Scientific Supplies
PGE075
470020-676EA 22.15 USD InStock
470020-676
Equilateral Prism
Educational Materials Physics Educational Materials Optics Learning Activities
A must for your optics lab
  • Durable
  • Glass
Teach students about the inner workings of the optical instruments they use and explore basic optical properties of light such as refraction, dispersion, and spectral phenomena.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty