Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Tree Calipers
Supplier: PECO SALES INC SE
59722
470021-026EA 374.75 USD InStock
470021-026
Tree Calipers
Calipers
Easily Take Accurate Measurements in the Field
Made of aluminum, this lightweight tree caliper is a valuable forestry tool designed for ease of use. Measure the diameter of tree trunks and other large objects up to 24 inches with the precision of 1/10 inch graduations marked on both sides. Size: 34" x 16".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty