Would you like to log in again?
LaMotte® Ammonia-Nitrogen Water Test Kit
Supplier: LaMotte
3304-02
470021-194KT 107 USD InStock
470021-194
LaMotte® Ammonia-Nitrogen Water Test Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Suitable for use in fresh, brackish and salt water.
  • Range: 0 to 2 ppm
  • Materials for 50 tests
Precalibrated test tubes and labware give fast, accurate results on-site, in the classroom, or in the lab. Reagents, labware, and step-by-step instructions are furnished in a compact, waterproof carrying case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty