Would you like to log in again?
Fieldmaster Turbidity Tube
Supplier: SCIENCE INTERACTIVE GROUP
623-2050
470021-244EA 109 USD InStock
470021-244
Fieldmaster Turbidity Tube
Tubes Test Tubes
Thsese tubes meets GLOBE requirements.
  • Clear polycarbonate construction
  • Secchi disk design on the bottom
  • Aligned with GLOBE protocols
Measure turbidity in streams where a Secchi disk cannot be used. Simply drain the tube with a hose and finger clamp.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty