Would you like to log in again?
Arsenic Quick Test Kit
Supplier: INDUSTRIAL TEST SYSTEMS INC SE
481396
470021-304EA 268.3 USD InStock
470021-304
Arsenic Quick Test Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Safe, Accurate, and Cost-Effective Test for Arsenic in Water or Soil
This colorimetric arsenic test only takes 12 minutes to complete, and it will detect a range of 0–500 ppb. The kit comes with 100 tests.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty