Would you like to log in again?
Chlorine/Ammonia Neutralizer
Supplier: PET WORLD INC SE
AMMOLOC45D
470021-340EA 21.3 USD InStock
470021-340
Chlorine/Ammonia Neutralizer
Specimens Live Specimens Aquaria - Living Material
Simplify aquarium setup.
  • Treats up to 480 gallons
  • Use for setup or water changes
Neutralize ammonia produced by fish waste, and remove chlorine and chloramine from tap water, with one easy-to-use treatment. It works in freshwater or saltwater aquariums.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty