Would you like to log in again?
Marina® Airline Attachments
Supplier: ROLF C. HAGEN (USA) CORP.
A1179 A1165 A1180 A1178 A1185
470308-744EA 3.5 USD InStock
470308-744 470308-746 470308-748 470005-156 470021-392
Marina® Airline Attachments
Animal Supplies Animal Environmental Enrichment
A necessity for any aquaria.
  • Use with fresh or salt water tanks
  • Gang valves fit onto the back of the tank
With a single pump, how are you going to run all of your fun decorations as well as your basic functions. Every aquaria needs some kind of airline attachments. The most common is a check valve to prevent an accidental syphoning back along the airline. You can easily attach multiple items to a single air pump with either a simple T-connector, or a gang valve. All plastic gang valves have a control knob, allowing you to control the amount of air traveling down each line.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty