Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Plane Mirror

Supplier: G.S.C. INTERNATIONAL, INC.

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

4-1301D
470021-852EA 5.1 USD InStock
470021-852
Plane Mirror
Mirrors
The silver-backed glass mirror measures 2×6", and has smooth ground edges.

The silver-backed glass mirror measures 2" x 6", and has smooth ground edges.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty