Would you like to log in again?
Insect Parts Microslide
Supplier: SUPERTEK SCIENTIFIC LLC
T-18-15B T-18
470022-058KT 191.25 USD InStock
470022-058 470008-992
Insect Parts Microslide
Slides Prepared Slides
This microslide illustrates insect anatomy up close.
  • Arrows Help Students Locate Important Features
  • Mounted in a Clear Plastic Holder That Protects It on Both Sides
  • Set of 15 Includes Detailed Lesson Plan on the Topic Under Study
  • Durable, Inexpensive Teaching Resource
Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides.
A detailed text folder is included.
Accessories information: Viewers are available separately.
Ordering information: Set includes Body of an insect (1X), Insect mouths (wm, 1X), Insect legs(wm, 1X), Honeybee leg (wm, 1X), Insect wings (1X), Antennae (wm, 1X), Compound eye (wm, 1X), and Breathing holes (1X).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty