Would you like to log in again?
Monocots and Dicots Microslide
Supplier: SUPERTEK SCIENTIFIC LLC
T-210-15-B T-210
470009-094PK 12.8 USD InStock
470009-094 470022-066
Monocots and Dicots Microslide
Slides Prepared Slides
Flowering Plant Comparisons
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: Set includes Monocot seed (ls, 20X), Dicot seed (ls, 40X), Petunia seedling (wm,10X), Monocot and dicot leaves (wm), Monocot and dicot roots (cs, 50X), Monocot and dicot stems (cs, 8X and 34X), Dicot stem; 3 yrs. old (wm,18X), and Monocot flower (cs, 6X).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty