Would you like to log in again?
Plant Mitosis Microslide, Set
Supplier: SUPERTEK SCIENTIFIC LLC
T-55-15B T-55
470022-102BX 191.75 USD InStock
470022-102 470009-020
Plant Mitosis Microslide, Set
These sets offer an alternate option for learning about the microscopic world without the use of a microscope.
  • Arrows Help Students Locate Important Features
  • Mounted in a Clear Plastic Holder That Protects It on Both Sides
  • Set of 15 Includes Detailed Lesson Plan on the Topic Under Study
  • Durable, Inexpensive Teaching Resource
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides.
A detailed text folder is included.
Ordering information: Set includes Early prophase (ls, st, 1,000X); Prophase (ls, st, 1,000X); Metaphase (ls, st, 1,000X); Early anaphase (ls, st, 1,000X); Anaphase (ls, st, 1,000X); Late anaphase (ls, st, 1,000X); Telophase (ls, st, 1,000X); and Late telophase (ls, st, 1,000X). Box of 15 sets also includes background subject information and reproducible student worksheets with answer keys.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty