Would you like to log in again?
The Circulatory System Microslide
Supplier: SUPERTEK SCIENTIFIC LLC
T-68 T-68-15B
470009-040PK 12.8 USD InStock
470009-040 470022-120
The Circulatory System Microslide
Slides Prepared Slides
Explore the pathways and tissues of the circulatory system.
  • Arrows Help Students Locate Important Features
  • Mounted in a Clear Plastic Holder That Protects It on Both Sides
  • Set of 15 Includes Detailed Lesson Plan on the Topic Under Study
  • Durable, Inexpensive Teaching Resource
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions.
You can supply all your students with their own microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: This set includes human blood (sm, st, 900X), phagocyte (em, 25,000X), artery and vein (cs, st, 35X), capillaries in the lung (cs, i, 360X), lymph vessel and valves (wm, st, 60X), rat heart (ls, st, 4X), rat heart (cs, st, 6X), and valve at aorta (ls, st, 7 1/2X).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty