Would you like to log in again?
Chromosomes and Genes in Action Microslide
Supplier: SUPERTEK SCIENTIFIC LLC
T-81-15B T-81
470022-136BX 192.15 USD InStock
470022-136 470009-056
Chromosomes and Genes in Action Microslide
Slides Prepared Slides
Investigate Genetic Material
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: Set includes DNA molecule (em, 55,000X), Coiling in chromosomes (wm, 2,000X), Lampbrush chromosomes (pc, 700X), Loop of lampbrush chromosomes (em, 9,000X/15,000X),
Giant chromosomes (wm, 1,000X), Chromosomes puffs, stained (250X), Chromosome puffs, tagged (250X), Nuclear pores (em, 50,000X), and Ribosomes (em, 85,000X).
Box of 15 Sets also includes Background subject information and Reproducible student worksheets with answer keys.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty