Would you like to log in again?
Viruses Microslide, Set
Supplier: SUPERTEK SCIENTIFIC LLC
T-97 T-97-15B
470022-148BX 192.15 USD InStock
470022-148 470009-070
Viruses Microslide, Set
Slides Prepared Slides
Delve into the world of microbes and viruses.
  • Arrows Help Students Locate Important Features
  • Mounted in a Clear Plastic Holder That Protects It on Both Sides
  • Set of 15 Includes Detailed Lesson Plan on the Topic Under Study
  • Durable, Inexpensive Teaching Resource
How small is a virus? Do viruses look like our cells or even like other viruses? Compare and contrast disease-causing and helpful viruses.
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides. A detailed text folder is included. Viewers are available separately.
Contents:
Lambda phage on E. coli (em, 45,000X)
Disrupted phage (em, 280,000X)
Human viruses:
Influenza virus (em, 170,000X) and Polio virus (em, 290,000X)
Measles virus attack (fs, 400)
Animal viruses:
Newcastle disease (em, 20,000X) and Vaccinia virus (em, 120,000X)
Viruses emerging from cell (em, 90,000X)
Cancer viruses:
Papilloma virus (em, 7,000X) and Rauscher leukemia virus (em, 90,000X)
Box of 15 Sets also Includes:
Background subject information
Reproducible student worksheets with answer keys
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty