Would you like to log in again?
Ward's® Slide Boxes
Supplier: Avantor
Ward's®
WARD470022-474
470022-478EA 6.95 USD InStock
470022-478 470022-476 470022-474
Ward's® Slide Boxes
Microscope Slide Boxes
Sturdy cardboard containers hold specimens securely.
  • Multiple sizes available
  • Display and storage purposes
Heavy-grade cardboard makes our collection boxes stronger than standard boxes. The clean white interiors and leatherette-style exteriors add distinction, while the telescoping lids fit snugly. Each box holds your choice of two compartmented trays, sold separately.
Small collection box: Holds 12-and 16-compartment storage trays. Size: 7¹/₄"L×7¹/₄"W×1¹/₂"H.
Medium collection box: Holds 24-and 36-compartment storage trays. Size: 13¹/₄"L×10¹/₄"W×1³/₄"H.
Large collection box: Holds 45-and 50-compartment storage trays. Size: 16¹/₂"L×13"W×1³/₄"H.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty