Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ward's® Plant And Animal Cell Structure Poster Sets
Supplier: Avantor
Ward's®
WARD470023-242
470023-242KT 64.99 USD InStock
470023-242
Ward's® Plant And Animal Cell Structure Poster Sets
Charts
Large Posters Display Intricate Features Of Cell In Vivid Color
  • Bright, easy-to-see illustrations
The illustrations of a typical plant cell or a typical animal cell have clearly labeled parts. The two charts also feature descriptions of the structure and function of each organelle and its relation to the entire cell structure. The plant cell poster shows the cell wall, large vacuole, chloroplasts, nucleus, ribosomes, Golgi complex, mitochondria, and more; the animal cell poster shows the plasma membrane, nucleus, ribosomes, Golgi complex, mitochondria, centrioles, cytoskeleton, and more.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty