Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ward's® Drosophila Life Cycle Poster
Supplier: Avantor
WARD470023-246
470023-246EA 39.39 USD InStock
470023-246
Ward's® Drosophila Life Cycle Poster
Charts
Magnified Photos Show Development and Traits of the Fruit Fly
  • A must-have reference for your Drosophila genetics lab
This large poster clearly illustrates the four stages in the life cycle of the fruit fly. Many different phenotypes commonly used for identification, such as eye color, wing type, bristles, and body color, are prominently visible in up-close photographs. The photos also show examples of sex combs and abdomen color, which are traits used for sex determination.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty