Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ward's® Blood Typing Poster
Supplier: Avantor
WARD470023-256
470023-256EA 37.95 USD InStock
470023-256
Ward's® Blood Typing Poster
Charts
This blood typing poster allows students to visualize microscopic blood components involved in the blood typing reaction.
Your students will understand the relationship between red blood cells, antigens, and antibodies, and how they interact in the determination of the four different blood types of the ABO system. Through the illustrations presented, students will begin to appreciate the importance of blood typing and how it relates to the outcome of a blood transfusion.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty