Would you like to log in again?
Introduction to Rocks Chart
Supplier: FISCHER TECHNICAL CO SE
E104L
470023-436EA 22.55 USD InStock
470023-436
Introduction to Rocks Chart
Charts
What Rocks Are and How They are Formed
Ideal for geology classes and collectors, this highly descriptive chart is also a helpful resource for identification. The chart explains how rocks are made of minerals and that each type of rock is the result of a specific mixture of minerals subjected to a clearly defined geological process. A section is dedicated to each type of rock — igneous, sedimentary, and metamorphic — with an illustration showing how they are made. A dramatic photograph also shows a natural formation, while additional photographs highlight a dozen representational specimens. A bonus section shows and explains some of the more specialized and popular rock forms such as geodes, concretions, agates, and thunder balls. Laminated. Size: 24" x 36".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty