Would you like to log in again?
Ward's® Exploring Density and Isostasy Lab Activity
Supplier: Avantor
WARD470023-706
470023-706KT 267.39 USD InStock
470023-706
Ward's® Exploring Density and Isostasy Lab Activity
Educational Classroom Kits and Activities
Re-create the Earth’s Crust Equilibrium
Students determine the density of three basic rock samples, relate them to the principle of isostasy, and examine how the Earth’s crust floats on the mantle by using blocks of wood and weights. The set includes a teacher’s guide and student copymasters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty