Would you like to log in again?
Lab-Aids® Nematode Study Kit
Supplier: LAB-AIDS INC (KITS) SE
34
470023-798EA 129 USD InStock
470023-798
Lab-Aids® Nematode Study Kit
Educational Materials Biology Educational Materials Animal Biology Learning Activites
Extract roundworms from soil for up-close study
  • Experience and study live, harmless nematodes
  • Explore characteristics and structure of nematodes
  • Develop microscope technique
  • Time required: two 50 minutes class period
This kit offers students a very effective means for learning about living organisms. Students extract nematodes and explore their characteristics and structure. Sampling is easy as Nematodes can be extracted from almost any type of soil sample. Everything needed to extract nematodes is included in this kit. It will require 24 hours to extract nematodes.
Delivery information: See datasheet for contents information.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty