Would you like to log in again?
Ward's® Genetic Test Paper Kit
Supplier: Avantor
WARD470023-856
470023-856KT 18.35 USD InStock
470023-856
Ward's® Genetic Test Paper Kit
Educational Classroom Kits and Activities
Examine a Genetically Inherited Trait in the Classroom
The ability or inability to taste certain chemicals is genetically predetermined. Students can test their own taste responses using the specially prepared test papers included in this kit. They can then pool the class data and apply the Hardy-Weinberg Principle to determine the frequency of dominant and recessive alleles within the population, consisting of the class. You will get PTC test paper, thiourea test paper, sodium benzoate test paper, and control test paper (100 strips of each), as well as a teacher’s guide and student copymaster.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty