Would you like to log in again?
Ward's® Nonsilicate Mineral Specimens Card Set
Supplier: Ward's
362501
470023-890KT 6.49 USD InStock
470023-890
Ward's® Nonsilicate Mineral Specimens Card Set
Educational Materials Earth Science Educational Materials Earth Science Textbooks
There’s no more convenient way of learning essential properties of minerals than with this informative, decorative set of cards exclusively from WARD’S.
  • Size: 3½"×2½
Vivid photos and important information on a variety of mineral types are all contained on these trading-card-size reference cards, making them useful as flash cards to help students learn or review key properties and features, or as informational supplements to specimen collections. The mineral specimen cards list the mineral group, chemical composition, crystallography, physical properties, composition, occurrence, and uses.
Mineral groups: native elements; sulfides and sulfosalts; oxides; hydroxides; halides; carbonates; borates, phosphates, and tungstates; and sulfates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty