Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Recyclable Plastics: Identification using Relative Density and Flammability
Supplier: LAB-AIDS INC (KITS) SE
704
470023-904EA 349.15 USD InStock
470023-904
Recyclable Plastics: Identification using Relative Density and Flammability
Educational Materials Biology Educational Materials Biochemistry Learning Activities
Identify and Construct Unknown Polymers
  • Denity, Polymers, Enviornmental Chemistry and More
Besides having students identify polymers, this lab activity can be used as an enrichment unit on density. Students use specially designed trays to identify recyclable plastics by determining their relative density. They also test the flammability of the plastics and their physical characteristics. Students attempt to identify unknown plastics using the information they have collected. Then, using molecular model components, students construct each repeat unit (the continuous molecule that makes up the larger polymer or plastic) in the polymer structure.
Ordering information: Includes Teacher Guide with SDS, Student Worksheets and Guides, Candles in holders, Containers with Lids, Copper Wires, Density Trays, Forceps, Stock Density solutions, Unknown plastic strips package, Resin package, Plastic strip package, and Molecular model component Package.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty