Would you like to log in again?
Bacterial Pollution of Water Lab Activity
Supplier: THE SCIENCE SOURCE SE
1400
470023-912EA 102 USD InStock
470023-912
Bacterial Pollution of Water Lab Activity
Educational Materials Environmental Educational Materials Environmental Learning Activities
Use the Exact Water-Testing Procedures Used by Health Authorities
  • Collect your own samples
  • Practice preparing microscope slides
  • Includes a teacher's guide
Students will collect water samples, extract a few drops, and add them to an agar medium. They are able to see and study the bacterial colonies that form as a result. The kit contains all materials needed for 15 setups, including a teacher’s manual and student instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty